Methotrexate hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Antifolan hydrate
, MTX hydrate
, (+)-Amethopterin hydrate
, L-4-Amino-N10-methylpteroylglutamic acid hydrate
, (4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzoyl)-L-glutamic acid hydrate(1:x)
, L-Amethopterin hydrate
, 4-Amino-10-methylfolic acid hydrate
, Methylaminopterin hydrate
, L(+)-Amethopterin hydrate
CAS Number: 59-05-2
MDL Number: MFCD00150847
Molecular Formula: C20H22N8O5
Molecular Weight: 454.45
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 200
Specific rotation [a]20/D: 21 deg (C=0.1, 1mol/L NaOH sol.)
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immunosuppressant Ingredients, Synthetic Antimicrobial Ingredients, Antitumor Ingredients, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredie
Other Product Names / Synonyms: (+)-Amethopterin
Formula:
C₂₀H₂₂N₈O₅ MW: 454.44 g/mol (anhy) Melting Pt: 200 °C |
MDL Number:
MFCD00150847 CAS Number: 59-05-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...