(S)-(-)-4-Methoxy-α-methylbenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-(-)-1-(4-Methoxyphenyl)ethylamine(S)-1-(4-Methoxyphenyl)ethylamine

M1512-25G M1512-5G
TCM1512-5GEA 88 USD
TCM1512-5G TCM1512-25G
(S)-(-)-4-Methoxy-α-methylbenzylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-4-Methoxy-α-methylbenzylamine

CAS Number: 41851-59-6
MDL Number: MFCD00671660
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.03
Specific rotation [a]20/D: -32 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-4-Methoxy-alpha-methylbenzylamine

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Density: 1.03 (20 °C)
MDL Number: MFCD00671660
CAS Number: 41851-59-6
UN: 2735
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/57/76/28975776.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR