7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidineMTBD

M1443-5G M1443-1G
TCM1443-5GEA 269 USD
TCM1443-5G TCM1443-1G
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene ≥95.0% (by GC, titration analysis)
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene

CAS Number: 84030-20-6
MDL Number: MFCD00043004
Molecular Formula: C8H15N3
Molecular Weight: 153.23
Purity/Analysis Method: 95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 113
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine, MTBD

Formula: C₈H₁₅N₃
MW: 153.23 g/mol
Boiling Pt: 79 °C
Density: 1.06 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00043004
CAS Number: 84030-20-6
https://us.vwr.com/stibo/web/std.lang.all/56/16/28975616.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR