ω-Bromo-4-methoxyacetophenone ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Bromo-1-(4-methoxyphenyl)ethan-1-one
CAS Number: 2632-13-5
MDL Number: MFCD00000201
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 71
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: alpha-Bromo-4'-methoxyacetophenone, 4-(Bromoacetyl)anisole
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Melting Pt: 71 °C |
MDL Number:
MFCD00000201 CAS Number: 2632-13-5 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...