Methyl dichlorophosphite ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Methyl phosphorodichloridite

M0905-10G
TCM0905-10GEA 92.37 USD
TCM0905-10G
Methyl dichlorophosphite ≥95.0% (by titrimetric analysis)
Methyl dichlorophosphite

[Phosphitylating Agent]
CAS Number: 3279-26-3
MDL Number: MFCD00000524
Molecular Formula: CH3Cl2OP
Molecular Weight: 132.91
Purity/Analysis Method: 95.0% (T)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 40
Specific Gravity (20/20): 1.38
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Methyl Phosphorodichloridite

Formula: CH₃Cl₂OP
MW: 132.91
Boiling Pt: 92 °C
Density: 1.38 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00000524
CAS Number: 3279-26-3
UN: 2920
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/44/37/28974437.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR