1-Methylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-methylcyclopropane-1-carboxylic acid
M0885-5G
M0885-25G
TCM0885-25GEA
567.22
USD
TCM0885-25G
TCM0885-5G
1-Methylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-Methylcyclopropanecarboxylic acid
CAS Number: 6914-76-7
MDL Number: MFCD00001290
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 185
Flash Point (°C): 84
Freezing point (°C): 33
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 185 °C Flash Pt: 84 °C |
MDL Number:
MFCD00001290 CAS Number: 6914-76-7 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...