2-Methoxypyridine ≥98.0% (by GC)

Supplier: TCI America

M0788-25ML
TCM0788-25MLEA 30 USD
TCM0788-25ML
2-Methoxypyridine ≥98.0% (by GC)
2-Methoxypyridine

CAS Number: 1628-89-3
MDL Number: MFCD00006262
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 32
Specific Gravity (20/20): 1.05

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 142 °C
Density: 1.05 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00006262
CAS Number: 1628-89-3
UN: 3271
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/42/56/28974256.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR