TCIAM0638

M0638-100MG
TCM0638-100MGEA 166.99 USD
TCM0638-100MG
TCIAM0638
CAS Number: 1039-17-4
MDL Number: MFCD00021151
Molecular Formula: C20H28O2
Molecular Weight: 300.44
Purity/Analysis Method: 90.0% (GC)
Form: Grain
Melting point (°C): 169

Product Categories: Bioscience, Steroids, Androstanes
Other Product Names / Synonyms: 17beta-Hydroxy-17-methylandrosta-4,9(11)-dien-3-one
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR