1-Methyl-2-thiourea ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Methylthiourea

M0619-25G
TCM0619-25GEA 55.83 USD
TCM0619-25G
1-Methyl-2-thiourea ≥98.0% (by titrimetric analysis)
1-Methyl-2-thiourea

CAS Number: 598-52-7
MDL Number: MFCD00004938
Molecular Formula: C2H6N2S
Molecular Weight: 90.14
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 120

Formula: C₂H₆N₂S
MW: 90.15 g/mol
Melting Pt: 120 °C
MDL Number: MFCD00004938
CAS Number: 598-52-7
UN: 2811
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/40/80/28974080.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR