1-Methyl-1-phenylhydrazine ≥95.0% (by GC)

Supplier: TCI America

M0399-5G M0399-25G
TCM0399-5GEA 83 USD
TCM0399-5G TCM0399-25G
1-Methyl-1-phenylhydrazine ≥95.0% (by GC)
1-Methyl-1-phenylhydrazine

CAS Number: 618-40-6
MDL Number: MFCD00007627
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 92
Specific Gravity (20/20): 1.04

Formula: C₇H₁₀N₂
Boiling Pt: 109 °C
Density: 1.04 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00007627
CAS Number: 618-40-6
https://us.vwr.com/stibo/web/std.lang.all/38/21/28973821.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR