4-Methyl-2-pentene ≥96.0% (by GC)

Supplier: TCI America

M0395-5ML
TCM0395-5MLEA 198.88 USD
TCM0395-5ML
4-Methyl-2-pentene ≥96.0% (by GC)
4-Methyl-2-pentene

CAS Number: 4461-48-7
MDL Number: MFCD00065138
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 58
Flash Point (°C): -32
Specific Gravity (20/20): 0.67

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 58 °C
Density: 0.67 (20 °C)
Flash Pt: –32 °C
MDL Number: MFCD00065138
CAS Number: 4461-48-7
https://us.vwr.com/stibo/web/std.lang.all/38/17/28973817.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR