Methylhydrazine sulfate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
M0341-5G
M0341-25G
TCM0341-5GEA
55.81
USD
TCM0341-5G
TCM0341-25G
Methylhydrazine sulfate ≥98.0% (by titrimetric analysis)
Methylhydrazine sulfate
CAS Number: 302-15-8
MDL Number: MFCD00035423
Molecular Formula: CH6N2
Molecular Weight: 144.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 143
Formula:
CH₆N₂ Melting Pt: 143 °C |
MDL Number:
MFCD00035423 CAS Number: 302-15-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...