5-Methyl-2-hexene (cis- and trans- mixture) ≥99.0% (by GC)

Supplier: TCI America

M0305-1ML
TCM0305-1MLEA 112.62 USD
TCM0305-1ML
5-Methyl-2-hexene (cis- and trans- mixture) ≥99.0% (by GC)
5-Methyl-2-hexene (cis- and trans- mixture)

CAS Number: 3404-62-4
MDL Number: MFCD00059239
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Flash Point (°C): -7
Specific Gravity (20/20): 0.70

Formula: C₇H₁₄
Density: 0.70 (20 °C)
Flash Pt: –7 °C
MDL Number: MFCD00059239
CAS Number: 3404-62-4
https://us.vwr.com/stibo/web/std.lang.all/37/09/28973709.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR