9-Methylanthracene ≥98.0% (by GC)

Supplier: TCI America

M0153-5G M0153-1G
TCM0153-1GEA 18 USD
TCM0153-1G TCM0153-5G
9-Methylanthracene ≥98.0% (by GC)
9-Methylanthracene

CAS Number: 779-02-2
MDL Number: MFCD00001261
Molecular Formula: C15H12
Molecular Weight: 192.26
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 370
Melting point (°C): 79

Formula: C₁₅H₁₂
MW: 192.26 g/mol
Boiling Pt: 370 °C
Melting Pt: 79 °C
MDL Number: MFCD00001261
CAS Number: 779-02-2
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/34/52/28973452.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR