Letrozole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-Bis(4-cyanophenyl)methyl-1,2,4-triazole4,4'-(1,2,4-Triazol-1-ylmethylene)dibenzonitrile

L0248-1G
TCL0248-1GEA 302.64 USD
TCL0248-1G
Letrozole ≥98.0% (by HPLC, titration analysis)
Letrozole

CAS Number: 112809-51-5
MDL Number: MFCD00866241
Molecular Formula: C17H11N5
Molecular Weight: 285.31
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 185
Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Inhibitors (for Pharmacological Research), Antitumor Ingredients
Other Product Names / Synonyms: 1-Bis(4-cyanophenyl)methyl-1,2,4-triazole, 4,4'-(1,2,4-Triazol-1-ylmethylene)dibenzonitrile

Formula: C₁₇H₁₁N₅
MW: 285.31 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00866241
CAS Number: 112809-51-5
https://us.vwr.com/stibo/web/std.lang.all/31/82/28973182.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR