2,3-Lutidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,3-Dimethylpyridine
L0063-25ML
TCL0063-25MLEA
25.35
USD
TCL0063-25ML
2,3-Lutidine ≥98.0% (by GC, titration analysis)
2,3-Lutidine
CAS Number: 583-61-9
MDL Number: MFCD00009605
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 161
Melting point (°C): -15
Flash Point (°C): 50
Specific Gravity (20/20): 0.95
Product Categories: Synthetic Organic Chemistry, Ligands, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 2,3-Dimethylpyridine
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 161 °C Melting Pt: –15 °C Density: 0.95 (20 °C) Flash Pt: 50 °C |
MDL Number:
MFCD00009605 CAS Number: 583-61-9 UN: 2929 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...