2-Iodoisophthalic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0883-5G
TCI0883-5GEA 299 USD
TCI0883-5G
2-Iodoisophthalic acid ≥98.0% (by GC, titration analysis)
2-Iodoisophthalic acid

CAS Number: 2902-65-0
MDL Number: MFCD00156976
Molecular Formula: C8H5IO4
Molecular Weight: 292.03
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 245

Formula: C₈H₅IO₄
MW: 292.03 g/mol
Melting Pt: 245 °C
MDL Number: MFCD00156976
CAS Number: 2902-65-0
https://us.vwr.com/stibo/web/std.lang.all/27/65/28972765.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR