2-Isopropylthiophenol ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 2-Isopropylthiophenol1-Isopropyl-2-mercaptobenzene2-Isopropylbenzenethiol2-Isopropylbenzene-1-thiol

I0857-5G
TCI0857-5GEA 321.76 USD
TCI0857-5G
2-Isopropylthiophenol ≥95.0% (by GC)
2-Isopropylthiophenol

CAS Number: 6262-87-9
MDL Number: MFCD00004837
Molecular Formula: C9H12S
Molecular Weight: 152.26
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 227
Flash Point (°C): 86
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: 2-Isopropylthiophenol

Formula: C₉H₁₂S
MW: 152.26 g/mol
Boiling Pt: 227 °C
Density: 1.01 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00004837
CAS Number: 6262-87-9
https://us.vwr.com/stibo/web/std.lang.all/27/45/28972745.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR