(S)-(-)-Sulpiride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (S)-N-[(1-Ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamideLevosulpiride

I0821-5G I0821-25G
TCI0821-5GEA 109.59 USD
TCI0821-5G TCI0821-25G
(S)-(-)-Sulpiride ≥98.0% (by HPLC, titration analysis)
(S)-(-)-Sulpiride

CAS Number: 23672-07-3
MDL Number: MFCD00055220
Molecular Formula: C15H23N3O4S
Molecular Weight: 341.43
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 187
Specific rotation [a]20/D: -69 deg (C=3, DMF)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (S)-N-[(1-Ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamide, Levosulpiride

Formula: C₁₅H₂₃N₃O₄S
MW: 341.43 g/mol
Melting Pt: 187 °C
MDL Number: MFCD00055220
CAS Number: 23672-07-3
https://us.vwr.com/stibo/web/std.lang.all/27/11/28972711.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR