(R,R)-2-Iodo-1,3-bis[1-(mesitylcarbamoyl)ethoxy]benzene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (R,R)-2-Iodo-1,3-bis[1-(2,4,6-trimethylphenylcarbamoyl)ethoxy]benzene

I0807-200MG
TCI0807-200MGEA 124.81 USD
TCI0807-200MG
(R,R)-2-Iodo-1,3-bis[1-(mesitylcarbamoyl)ethoxy]benzene ≥98.0% (by HPLC)
(R,R)-2-Iodo-1,3-bis[1-(mesitylcarbamoyl)ethoxy]benzene

CAS Number: 1226896-38-3
Molecular Formula: C30H35IN2O4
Molecular Weight: 614.52
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (R,R)-2-Iodo-1,3-bis[1-(2,4,6-trimethylphenylcarbamoyl)ethoxy]benzene

Formula: C₃₀H₃₅IN₂O₄
MW: 614.52 g/mol
CAS Number: 1226896-38-3
https://us.vwr.com/stibo/web/std.lang.all/26/94/28972694.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR