(R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(R,R)-(+)-2,2'-(Dimethylmethylene)bis(4-tert-butyl-2-oxazoline)
CAS Number: 131833-97-1
MDL Number: MFCD07368371
Molecular Formula: C17H30N2O2
Molecular Weight: 294.44
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 90
Specific rotation [a]20/D: 119 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (R,R)-(+)-2,2'-(Dimethylmethylene)bis(4-tert-butyl-2-oxazoline), (R,R)-(+)-2,2-Bis(4-tert-butyl-2-oxazolin-2-yl)propane
Formula:
C₁₇H₃₀N₂O₂ MW: 294.44 g/mol Melting Pt: 90 °C |
MDL Number:
MFCD07368371 CAS Number: 131833-97-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...