2-Iodoxybenzoic acid 39.0% (by titrimetric analysis) stabilized

Supplier: TCI America

Synonyms: 2-Iodylbenzoic acid1-Hydroxy-1,2-benziodoxy-3-(1h)-one-1-xoideIBX

I0791-25G I0791-5G
TCI0791-5GEA 92.65 USD
TCI0791-5G TCI0791-25G
2-Iodoxybenzoic acid 39.0% (by titrimetric analysis) stabilized
2-Iodoxybenzoic acid

(stabilized with Benzoic Acid + Isophthalic Acid)
CAS Number: 61717-82-6
MDL Number: MFCD02912492
Molecular Formula: C7H5IO4
Molecular Weight: 280.02
Purity/Analysis Method: 39.0% (T)
Form: Crystal
Melting point (°C): 232


Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Oxidation
Other Product Names / Synonyms: IBX

Formula: C₇H₅IO₄
MW: 280.02 g/mol
Melting Pt: 232 °C
MDL Number: MFCD02912492
CAS Number: 61717-82-6
https://us.vwr.com/stibo/web/std.lang.all/26/77/28972677.jpg

Order Now

Specification Test Results

Purity/Analysis Method 39.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR