4-Iodobiphenyl ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4-Iodo-1,1'-biphenyl

I0785-5G I0785-25G
TCI0785-25GEA 109.62 USD
TCI0785-25G TCI0785-5G
4-Iodobiphenyl ≥97.0% (by GC)
4-Iodobiphenyl

CAS Number: 1591-31-7
MDL Number: MFCD00019028
Molecular Formula: C12H9I
Molecular Weight: 280.11
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 320
Melting point (°C): 112

Formula: C₁₂H₉I
MW: 280.11 g/mol
Boiling Pt: 320 °C
Melting Pt: 112 °C
MDL Number: MFCD00019028
CAS Number: 1591-31-7
https://us.vwr.com/stibo/web/std.lang.all/26/70/28972670.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR