3-(Isopropylamino)propanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(Isopropylamino)propan-1-ol

I0755-25G I0755-5G
TCI0755-25GEA 294.26 USD
TCI0755-25G TCI0755-5G
3-(Isopropylamino)propanol ≥98.0% (by GC, titration analysis)
3-(Isopropylamino)propanol

CAS Number: 33918-15-9
MDL Number: MFCD00483086
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 94
Freezing point (°C): 35

Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 94 °C
MDL Number: MFCD00483086
CAS Number: 33918-15-9
https://us.vwr.com/stibo/web/std.lang.all/26/36/28972636.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR