(Indenyl)titanium(IV) trichloride ≥98.0% (by gravimetric and titrimetric analysis)

Supplier: TCI America

Synonyms: Trichloro(indenyl)titanium(IV)

I0646-1G
TCI0646-1GEA 634.08 USD
TCI0646-1G
(Indenyl)titanium(IV) trichloride ≥98.0% (by gravimetric and titrimetric analysis)
(Indenyl)titanium(IV) trichloride

CAS Number: 84365-55-9
Molecular Formula: C9H7Cl3Ti
Molecular Weight: 269.37
Purity/Analysis Method: 98.0% (W,T)
Form: Crystal
Melting point (°C): 162


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: Trichloro(indenyl)titanium(IV)

Formula: C₉H₇Cl₃Ti
MW: 269.37 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00269849
CAS Number: 84365-55-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (W,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR