2-Isopropyl-2-oxazoline ≥97.0% (by GC)

Supplier: TCI America

I0642-5G
TCI0642-5GEA 247.5 USD
TCI0642-5G
2-Isopropyl-2-oxazoline ≥97.0% (by GC)
2-Isopropyl-2-oxazoline

CAS Number: 10431-99-9
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Specific Gravity (20/20): 0.95

https://us.vwr.com/stibo/web/std.lang.all/25/23/28972523.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR