4-Isopropylthiophenol ≥94.0% (by GC)

Supplier: TCI America

I0596-25G
TCI0596-25GEA 101.5 USD
TCI0596-25G
4-Isopropylthiophenol ≥94.0% (by GC)
4-Isopropylthiophenol

CAS Number: 4946-14-9
MDL Number: MFCD00041505
Molecular Formula: C9H12S
Molecular Weight: 152.26
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 4-Isopropylthiophenol

Formula: C₉H₁₂S
MW: 152.26 g/mol
Boiling Pt: 105 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00041505
CAS Number: 4946-14-9
https://us.vwr.com/stibo/web/std.lang.all/24/91/28972491.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR