3,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,2-Dimethoxy-4-isothiocyanatobenzene3,4-Dimethoxyphenylisothiocyanate

I0541-5G I0541-25G
TCI0541-5GEA 61.92 USD
TCI0541-5G TCI0541-25G
3,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by titrimetric analysis)
3,4-Dimethoxyphenyl isothiocyanate

CAS Number: 33904-04-0
MDL Number: MFCD00041075
Molecular Formula: C9H9NO2S
Molecular Weight: 195.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 48


Other Product Names / Synonyms: Isothiocyanic Acid 3,4-Dimethoxyphenyl Ester

Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Melting Pt: 48 °C
MDL Number: MFCD00041075
CAS Number: 33904-04-0
UN: 2923
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/24/08/28972408.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR