4-Methoxyphenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America

I0513-5G
TCI0513-5GEA 68.01 USD
TCI0513-5G
4-Methoxyphenyl isothiocyanate ≥98.0% (by GC)
4-Methoxyphenyl isothiocyanate

CAS Number: 2284-20-0
MDL Number: MFCD00011676
Molecular Formula: C8H7NOS
Molecular Weight: 165.21
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 281
Melting point (°C): 18
Flash Point (°C): 110
Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: Isothiocyanic Acid 4-Methoxyphenyl Ester

Formula: C₈H₇NOS
MW: 165.22 g/mol
Boiling Pt: 281 °C
Melting Pt: 18 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00011676
CAS Number: 2284-20-0
https://us.vwr.com/stibo/web/std.lang.all/23/75/28972375.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR