Iodoacetonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Iodoacetonitrile

I0419-25G
TCI0419-25GEA 595.81 USD
TCI0419-25G
Iodoacetonitrile ≥98.0% (by GC)
Iodoacetonitrile

CAS Number: 624-75-9
MDL Number: MFCD00001886
Molecular Formula: C2H2IN
Molecular Weight: 166.95
Purity/Analysis Method: 98.0% (GC)
Form: Liquid
Boiling point (°C): 78
Flash Point (°C): 86
Specific Gravity (20/20): 2.31

Formula: C₂H₂IN
MW: 166.95 g/mol
Boiling Pt: 78 °C
Density: 2.31 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00001886
CAS Number: 624-75-9
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/22/60/28972260.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR