Ethyl isocyanatoacetate ≥97.0% (by GC)
Supplier: TCI America
I0386-25G
I0386-5G
TCI0386-25GEA
191.77
USD
TCI0386-25G
TCI0386-5G
Ethyl isocyanatoacetate ≥97.0% (by GC)
Ethyl isocyanatoacetate
CAS Number: 2949-22-6
MDL Number: MFCD00002040
Molecular Formula: C5H7NO3
Molecular Weight: 129.12
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 72
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: Isocyanatoacetic Acid Ethyl Ester
Formula:
C₅H₇NO₃ Boiling Pt: 80 °C Density: 1.13 (20 °C) Flash Pt: 72 °C |
UN:
2206 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...