α,α,α-Trifluoro-3-iodotoluene ≥97.0% (by GC) stabilized

Supplier: TCI America

Synonyms: 1-Iodo-3-(trifluoromethyl)benzene 3-Iodobenzotrifluoride α,α,α-Trifluoro-m-iodotoluene

I0385-25G
TCI0385-25GEA 28.42 USD
TCI0385-25G
α,α,α-Trifluoro-3-iodotoluene ≥97.0% (by GC) stabilized
α,α,α-Trifluoro-3-iodotoluene

(stabilized with Copper chip)
CAS Number: 401-81-0
MDL Number: MFCD00001049
Molecular Formula: C7H4F3I
Molecular Weight: 272.01
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 70
Specific Gravity (20/20): 1.88


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Iodo-3-(trifluoromethyl)benzene, 3-Iodo-alpha,alpha,alpha-trifluorotoluene

Formula: C₇H₄F₃I
MW: 272.01 g/mol
Boiling Pt: 183 °C
Density: 1.88 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00001049
CAS Number: 401-81-0
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/22/15/28972215.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR