2'-Hydroxy-5'-methylacetanilide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
2-Acetamido-4-methylphenol
H1442-5G
H1442-25G
TCH1442-25GEA
75.08
USD
TCH1442-25G
TCH1442-5G
2'-Hydroxy-5'-methylacetanilide ≥98.0% (by HPLC, total nitrogen)
2'-Hydroxy-5'-methylacetanilide
CAS Number: 6375-17-3
MDL Number: MFCD00020183
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 160
Other Product Names / Synonyms: 2-Acetamido-p-cresol, 2-Acetamido-4-methylphenol
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...