Hexaphenylbenzene
Supplier: TCI America
Synonyms:
3',4',5',6'-Tetraphenyl-o-terphenyl
H1412-1G
H1412-5G
TCH1412-1GEA
71.37
USD
TCH1412-1G
TCH1412-5G
Hexaphenylbenzene
Hexaphenylbenzene
CAS Number: 992-04-1
MDL Number: MFCD00003057
Molecular Formula: C42H30
Molecular Weight: 534.70
Form: Crystal
Color: White
Melting point (°C): 455
Formula:
C₄₂H₃₀ MW: 534.7 g/mol Melting Pt: 455 °C |
MDL Number:
MFCD00003057 CAS Number: 992-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...