6-Hydroxy-5-nitronicotinic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

H1410-1G H1410-5G
TCH1410-1GEA 117 USD
TCH1410-1G TCH1410-5G
6-Hydroxy-5-nitronicotinic acid ≥97.0% (by GC, titration analysis)
6-Hydroxy-5-nitronicotinic acid

CAS Number: 6635-31-0
MDL Number: MFCD02180840
Molecular Formula: C6H4N2O5
Molecular Weight: 184.11
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 291


Other Product Names / Synonyms: 6-Hydroxy-5-nitro-3-pyridinecarboxylic Acid

Formula: C₆H₄N₂O₅
MW: 184.1100 g/mol
Melting Pt: 291 °C
MDL Number: MFCD02180840
CAS Number: 6635-31-0
https://us.vwr.com/stibo/web/std.lang.all/14/93/28971493.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR