9-Heptadecanol ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Heptadecan-9-ol

H1401-1G
TCH1401-1GEA 122.75 USD
TCH1401-1G
9-Heptadecanol ≥95.0% (by GC)
9-Heptadecanol

CAS Number: 624-08-8
MDL Number: MFCD00046734
Molecular Formula: C17H36O
Molecular Weight: 256.47
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 60

Formula: C₁₇H₃₆O
MW: 256.47 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00046734
CAS Number: 624-08-8
https://us.vwr.com/stibo/web/std.lang.all/14/78/28971478.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR