2-Iodo-3-pyridinol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Hydroxy-2-iodopyridine

H1396-1G
TCH1396-1GEA 86 USD
TCH1396-1G
2-Iodo-3-pyridinol ≥98.0% (by GC)
2-Iodo-3-pyridinol

CAS Number: 40263-57-8
MDL Number: MFCD00023421
Molecular Formula: C5H4INO
Molecular Weight: 221.00
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 189


Other Product Names / Synonyms: 2-Iodo-3-pyridinol

https://us.vwr.com/stibo/web/std.lang.all/14/74/28971474.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR