L(+)-α-(4-Hydroxyphenyl)glycine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: H-Phg(4-OH)-OH

H1389-25G H1389-5G
TCH1389-25GEA 294.8 USD
TCH1389-25G TCH1389-5G
L(+)-α-(4-Hydroxyphenyl)glycine ≥99.0% (by HPLC, titration analysis)
L(+)-α-(4-Hydroxyphenyl)glycine

CAS Number: 32462-30-9
MDL Number: MFCD00065932
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: 99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 214
Specific rotation [a]20/D: 155 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-(+)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, L-(+)-2-(4-Hydroxyphenyl)glycine, Oxfenicine, H-Phg(4-OH)-OH

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00065932
CAS Number: 32462-30-9
https://us.vwr.com/stibo/web/std.lang.all/14/69/28971469.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR