(4-Hydroxy-3-nitrophenyl)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(4-Hydroxy-3-nitrophenyl)acetic acid

H1362-1G H1362-5G
TCH1362-1GEA 42.61 USD
TCH1362-1G TCH1362-5G
(4-Hydroxy-3-nitrophenyl)acetic acid ≥98.0% (by titrimetric analysis)
(4-Hydroxy-3-nitrophenyl)acetic acid

CAS Number: 10463-20-4
MDL Number: MFCD00007122
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 146
Storage Temperature: 0°C

Formula: (C₆H₃(OH)(NO₂))CH₂COOH
MW: 197.15 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00007122
CAS Number: 10463-20-4
https://us.vwr.com/stibo/web/std.lang.all/14/21/28971421.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR