6-Hydroxyindazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1359-1G
TCH1359-1GEA 84.21 USD
TCH1359-1G
6-Hydroxyindazole ≥98.0% (by GC, titration analysis)
6-Hydroxyindazole

CAS Number: 23244-88-4
MDL Number: MFCD00022788
Molecular Formula: C7H6N2O
Molecular Weight: 134.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 219

Formula: C₇H₆N₂O
MW: 134.1353 g/mol
Melting Pt: 219 °C
MDL Number: MFCD00022788
CAS Number: 23244-88-4
https://us.vwr.com/stibo/web/std.lang.all/14/18/28971418.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR