N-(3-Hydroxypropyl)morpholine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-(4-Morpholinyl)-1-propanol3-Morpholino-1-propanol3-Morpholinopropanol4-(3-Hydroxypropyl)morpholine

H1355-25G
TCH1355-25GEA 110.64 USD
TCH1355-25G
N-(3-Hydroxypropyl)morpholine ≥98.0% (by GC)
N-(3-Hydroxypropyl)morpholine

CAS Number: 4441-30-9
MDL Number: MFCD01713533
Molecular Formula: C7H15NO2
Molecular Weight: 145.20
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 241
Flash Point (°C): 85
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 3-Morpholinopropanol, 3-Morpholino-1-propanol

https://us.vwr.com/stibo/web/std.lang.all/14/03/28971403.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR