4-(2-(Dimethylamino)ethyl)phenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Hordenine
H1351-1G
H1351-5G
TCH1351-5GEA
111.65
USD
TCH1351-5G
TCH1351-1G
4-(2-(Dimethylamino)ethyl)phenol ≥98.0% (by GC, titration analysis)
4-(2-(Dimethylamino)ethyl)phenol
CAS Number: 539-15-1
MDL Number: MFCD00051462
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 118
Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: Anhaline, 4-[2-(Dimethylamino)ethyl]phenol, 4-Hydroxy-N,N-dimethylphenethylamine
Formula:
C₁₀H₁₅NO Boiling Pt: 174 °C Melting Pt: 118 °C |
MDL Number:
MFCD00051462 CAS Number: 539-15-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...