(S)-2-Hydroxy-4-phenylbutyric acid ≥98.0% (by GC, titration analysis)

H1339-1G
TCH1339-1GEA 35.02 USD
TCH1339-1G
(S)-2-Hydroxy-4-phenylbutyric acid ≥98.0% (by GC, titration analysis)
(S)-2-Hydroxy-4-phenylbutyric acid

CAS Number: 115016-95-0
MDL Number: MFCD06656492
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 117
Specific rotation [a]20/D: 9 deg (C=1, EtOH)

Formula: C10H12O3
MW: 180.2005 g/mol
Melting Pt: 117 °C
MDL Number: MFCD06656492
CAS Number: 115016-95-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR