(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(+/-)-cis-Hexahydro-7a-hydroxy-3a-methyl-1H-indene-1,4(2H)-dione
H1318-50MG
TCH1318-50MGEA
581
USD
TCH1318-50MG
(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione ≥98.0% (by GC)
(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione
CAS Number: 1192178-33-8
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 101
Other Product Names / Synonyms: (+/-)-cis-Hexahydro-7a-hydroxy-3a-methyl-1H-indene-1,4(2H)-dione
Formula:
C₁₀H₁₄O₃ MW: 182.22 g/mol Melting Pt: 101 °C |
CAS Number:
1192178-33-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...