2'-Hydroxy-3-phenylpropiophenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(2-Hydroxyphenyl)-3-phenylpropan-1-one

H1313-25G
TCH1313-25GEA 168 USD
TCH1313-25G
2'-Hydroxy-3-phenylpropiophenone ≥98.0% (by GC)
2'-Hydroxy-3-phenylpropiophenone

CAS Number: 3516-95-8
MDL Number: MFCD00002221
Molecular Formula: C15H14O2
Molecular Weight: 226.28
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 30
Flash Point (°C): 113
Lambda max.: 324 nm (MeOH)


Other Product Names / Synonyms: 2-Hydroxyphenyl Phenethyl Ketone, 2-Hydroxyphenyl 2-Phenylethyl Ketone

Formula: C₁₅H₁₄O₂
MW: 226.2705 g/mol
Boiling Pt: 158 °C
Melting Pt: 30 °C
Flash Pt: 113 °C
MDL Number: MFCD00002221
CAS Number: 3516-95-8
https://us.vwr.com/stibo/web/std.lang.all/12/92/28971292.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR