Hexamethylditin ≥98.0% (by GC)
Supplier: TCI America
H1312-5G
H1312-1G
TCH1312-5GEA
116.86
USD
TCH1312-5G
TCH1312-1G
Hexamethylditin ≥98.0% (by GC)
Hexamethylditin
CAS Number: 661-69-8
MDL Number: MFCD00008277
Molecular Formula: C6H18Sn2
Molecular Weight: 327.63
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 182
Flash Point (°C): 61
Freezing point (°C): 22
Other Product Names / Synonyms: Hexamethyldistannane, Bis(trimethyltin)
Formula:
C₆H₁₈Sn₂ MW: 327.63 g/mol Boiling Pt: 182 °C Flash Pt: 61 °C |
MDL Number:
MFCD00008277 CAS Number: 661-69-8 UN: 2788 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...