(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol
, 2-(1-Hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile
H1303-1G
H1303-5G
TCH1303-5GEA
359.47
USD
TCH1303-5G
TCH1303-1G
(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile ≥98.0% (by HPLC, total nitrogen)
(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile
CAS Number: 93413-76-4
MDL Number: MFCD06658142
Molecular Formula: C15H19NO2
Molecular Weight: 245.32
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 127
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol
Formula:
C₁₅H₁₉NO₂ MW: 245.32 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD06658142 CAS Number: 93413-76-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...