5-(4-Hydroxybenzyl)-2,4-thiazolidinedione ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

H1299-100MG
TCH1299-100MGEA 85.17 USD
TCH1299-100MG
5-(4-Hydroxybenzyl)-2,4-thiazolidinedione ≥95.0% (by HPLC, titration analysis)
5-(4-Hydroxybenzyl)-2,4-thiazolidinedione

CAS Number: 74772-78-4
MDL Number: MFCD00754499
Molecular Formula: C10H9NO3S
Molecular Weight: 223.25
Purity/Analysis Method: 95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 157

Formula: C₁₀H₉NO₃S
Melting Pt: 157 °C
MDL Number: MFCD00754499
CAS Number: 74772-78-4
https://us.vwr.com/stibo/web/std.lang.all/12/58/28971258.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR