Hexadecanal ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Hexadecylaldehyde
H1296-1G
TCH1296-1GEA
400.8
USD
TCH1296-1G
Hexadecanal ≥97.0% (by GC)
Hexadecanal
CAS Number: 629-80-1
MDL Number: MFCD00055568
Molecular Formula: C16H32O
Molecular Weight: 240.43
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 151
Melting point (°C): 35
Storage Temperature:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Hexadecanaldehyde, Hexadecyl Aldehyde, Palmitaldehyde, Palmityl Aldehyde
Formula:
C₁₆H₃₂O MW: 240.43 g/mol Boiling Pt: 151 °C Melting Pt: 35 °C |
MDL Number:
MFCD00055568 CAS Number: 629-80-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...