Heptadecanal ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Heptadecylaldehyde

H1295-1G
TCH1295-1GEA 471.2 USD
TCH1295-1G
Heptadecanal ≥97.0% (by GC)
Heptadecanal

CAS Number: 629-90-3
MDL Number: MFCD04034658
Molecular Formula: C17H34O
Molecular Weight: 254.46
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 318
Melting point (°C): 34
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Heptadecanaldehyde, Heptadecyl Aldehyde

Formula: C₁₇H₃₄O
MW: 254.46 g/mol
Boiling Pt: 318 °C
Melting Pt: 34 °C
MDL Number: MFCD04034658
CAS Number: 629-90-3
https://us.vwr.com/stibo/web/std.lang.all/12/51/28971251.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR